Search
Search

Fubp1| Gene Symbol: Fubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841401.2 | XP_004841458.1 | 13 - 14 | 1171 | 77 | |
| XM_021255482.1 | XP_021111141.1 | 13 - 14 | 1171 | 77 | ||
| XM_021255483.1 | XP_021111142.1 | 13 - 14 | 1171 | 77 | ||
| XM_021255484.1 | XP_021111143.1 | 13 - 14 | 1168 | 77 | ||
| XM_021255485.1 | XP_021111144.1 | 13 - 14 | 1171 | 77 | ||
| XM_021255486.1 | XP_021111145.1 | 13 - 14 | 1171 | 77 | ||
| XM_021255487.1 | XP_021111146.1 | 12 - 13 | 1108 | 77 | ||
| XM_021255488.1 | XP_021111147.1 | 12 - 13 | 1105 | 77 | ||
| XM_021255489.1 | XP_021111148.1 | 12 - 13 | 1108 | 77 | ||
| XM_021255490.1 | XP_021111149.1 | 12 - 13 | 1105 | 77 | ||
| XM_021255491.1 | XP_021111150.1 | 12 - 13 | 1108 | 77 | ||
Set Membership: |
|