Search
Search

Sgk1| Gene Symbol: Sgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874826.3 | XP_004874883.1 | 6 - 7 | 1036 | 99 | |
| XM_004874827.3 | XP_004874884.1 | 4 - 5 | 409 | 99 | ||
| XM_004874828.2 | XP_004874885.1 | 4 - 5 | 442 | 99 | ||
| XM_013069284.2 | XP_012924738.1 | 4 - 5 | 414 | 99 | ||
| XM_021252194.1 | XP_021107853.1 | 5 - 6 | 541 | 99 | ||
Set Membership: |
|