Search
Search

Dcaf8| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858720.3 | XP_004858777.1 | 8 - 9 | 1423 | 97 | |
| XM_021239741.1 | XP_021095400.1 | 8 - 9 | 1538 | 97 | ||
| XM_021239742.1 | XP_021095401.1 | 9 - 10 | 1476 | 97 | ||
| XM_021239743.1 | XP_021095402.1 | 8 - 9 | 1374 | 97 | ||
| XM_021239744.1 | XP_021095403.1 | 8 - 9 | 1240 | 97 | ||
| XM_021239745.1 | XP_021095404.1 | 7 - 8 | 1326 | 97 | ||
| XM_021239746.1 | XP_021095405.1 | 8 - 9 | 1537 | 97 | ||
| XM_021239747.1 | XP_021095406.1 | 8 - 9 | 1536 | 97 | ||
| XM_021239748.1 | XP_021095407.1 | 8 - 9 | 1401 | 97 | ||
| XM_021239749.1 | XP_021095408.1 | 7 - 8 | 1310 | 97 | ||
Set Membership: |
|