Search
Search

Mgat5| Gene Symbol: Mgat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836099.3 | XP_004836156.1 | 12 - 13 | 1679 | 92 | |
| XM_004836100.3 | XP_004836157.1 | 12 - 13 | 1728 | 92 | ||
| XM_004836101.3 | XP_004836158.1 | 11 - 12 | 1660 | 92 | ||
| XM_021236863.1 | XP_021092522.1 | 12 - 13 | 1712 | 92 | ||
| XM_021236867.1 | XP_021092526.1 | 12 - 13 | 1644 | 92 | ||
| XM_021236872.1 | XP_021092531.1 | 12 - 13 | 1704 | 92 | ||
| XM_021236875.1 | XP_021092534.1 | 11 - 12 | 1639 | 92 | ||
| XM_021236883.1 | XP_021092542.1 | 11 - 12 | 1536 | 92 | ||
| XM_021236885.1 | XP_021092544.1 | 12 - 13 | 1732 | 92 | ||
Set Membership: |
|