Search
Search

Kifc3| Gene Symbol: Kifc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071602.2 | XP_012927056.1 | 16 - 17 | 2368 | 62 | |
| XM_013071604.2 | XP_012927058.1 | 15 - 16 | 2160 | 62 | ||
| XM_013071606.2 | XP_012927060.1 | 15 - 16 | 2276 | 62 | ||
| XM_021256986.1 | XP_021112645.1 | 16 - 17 | 2423 | 62 | ||
| XM_021256987.1 | XP_021112646.1 | 16 - 17 | 2423 | 62 | ||
| XM_021256988.1 | XP_021112647.1 | 16 - 17 | 2422 | 62 | ||
| XM_021256989.1 | XP_021112648.1 | 16 - 17 | 2422 | 62 | ||
| XM_021256990.1 | XP_021112649.1 | 14 - 15 | 2081 | 62 | ||
| XM_021256991.1 | XP_021112650.1 | 14 - 15 | 2080 | 62 | ||
| XM_021256992.1 | XP_021112651.1 | 16 - 17 | 2244 | 62 | ||
Set Membership: |
|