Search
Search

Stard3nl| Gene Symbol: Stard3nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840275.3 | XP_004840332.1 | 3 - 4 | 523 | 84 | |
| XM_013070557.2 | XP_012926011.1 | 3 - 4 | 609 | 84 | ||
| XM_013070558.2 | XP_012926012.1 | 3 - 4 | 507 | 84 | ||
| XM_021254563.1 | XP_021110222.1 | 3 - 4 | 524 | 84 | ||