Search
Search

LOC101701685| Gene Symbol: LOC101701685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842151.3 | XP_004842208.1 | 2 - 3 | 112 | 98 | |
| XM_021256095.1 | XP_021111754.1 | 2 - 3 | 281 | 98 | ||
Set Membership: |
|