Search
Search

Slc17a3| Gene Symbol: Slc17a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847579.3 | XP_004847636.2 | 2 - 3 | 494 | 59 | |
| XM_021260524.1 | XP_021116183.1 | 3 - 4 | 511 | 59 | ||
| XM_021260525.1 | XP_021116184.1 | 4 - 5 | 561 | 59 | ||
Set Membership: |
|