Search
Search

Grhl3| Gene Symbol: Grhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850591.2 | XP_004850648.1 | 12 - 13 | 1676 | 54 | |
| XM_021263069.1 | XP_021118728.1 | 12 - 13 | 1697 | 54 | ||
Set Membership: |
|