Search
Search

Cpsf4| Gene Symbol: Cpsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840511.2 | XP_004840568.1 | 5 - 6 | 595 | 57 | |
| XM_004840514.2 | XP_004840571.1 | 5 - 6 | 595 | 57 | ||
| XM_004840515.2 | XP_004840572.1 | 5 - 6 | 595 | 57 | ||
| XM_021254841.1 | XP_021110500.1 | 5 - 6 | 595 | 57 | ||
| XM_021254842.1 | XP_021110501.1 | 5 - 6 | 595 | 57 | ||
Set Membership: |
|