Search
Search

Cept1| Gene Symbol: Cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853698.3 | XP_004853755.1 | 2 - 3 | 535 | 69 | |
| XM_004853699.3 | XP_004853756.1 | 2 - 3 | 535 | 69 | ||
| XM_004853701.2 | XP_004853758.1 | 4 - 5 | 1320 | 69 | ||
| XM_021265642.1 | XP_021121301.1 | 3 - 4 | 1260 | 69 | ||
| XM_021265643.1 | XP_021121302.1 | 3 - 4 | 1257 | 69 | ||
Set Membership: |
|