Search
Search

Bbs1| Gene Symbol: Bbs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852298.2 | XP_004852355.1 | 11 - 12 | 1164 | 60 | |
| XM_021264542.1 | XP_021120201.1 | 11 - 12 | 1164 | 60 | ||
| XM_021264543.1 | XP_021120202.1 | 11 - 12 | 1127 | 60 | ||
Set Membership: |
|