Search
Search

Poglut1| Gene Symbol: Poglut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835672.2 | XP_004835729.1 | 4 - 5 | 558 | 73 | |
| XM_013075217.2 | XP_012930671.1 | 3 - 4 | 414 | 73 | ||
| XM_021263230.1 | XP_021118889.1 | 4 - 5 | 513 | 73 | ||
Set Membership: |
|