Search
Search

Riiad1| Gene Symbol: Riiad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854094.3 | XP_004854151.1 | 2 - 3 | 178 | 70 | |
| XM_013075450.2 | XP_012930904.1 | 2 - 3 | 178 | 70 | ||
Set Membership: |
|