Search
Search

Uap1| Gene Symbol: Uap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865166.2 | XP_004865223.1 | 6 - 7 | 1249 | 79 | |
| XM_004865167.3 | XP_004865224.1 | 6 - 7 | 1180 | 79 | ||
| XM_004865168.3 | XP_004865225.1 | 6 - 7 | 1249 | 79 | ||
| XM_021244649.1 | XP_021100308.1 | 6 - 7 | 1231 | 79 | ||
| XM_021244650.1 | XP_021100309.1 | 5 - 6 | 1018 | 79 | ||
Set Membership: |
|