Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hg05095757_m1
See other PARD3 GE Assays ›
Gene Symbol:
Pard3
Gene Name:
par-3 family cell polarity regulator
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624805: 7205028 - 7810050 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID
Hg05095757_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Pard3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_004861860.3
XP_004861917.1
12 - 13
1699
67
XM_004861861.3
XP_004861918.1
12 - 13
1699
67
XM_004861863.3
XP_004861920.1
12 - 13
1699
67
XM_004861865.2
XP_004861922.1
11 - 12
1790
67
XM_004861866.3
XP_004861923.1
12 - 13
1699
67
XM_004861867.3
XP_004861924.1
12 - 13
1873
67
XM_004861873.3
XP_004861930.1
12 - 13
1868
67
XM_004861874.2
XP_004861931.1
12 - 13
1864
67
XM_004861877.3
XP_004861934.1
12 - 13
1865
67
XM_013078500.2
XP_012933954.1
12 - 13
1874
67
XM_013078501.2
XP_012933955.1
12 - 13
1873
67
XM_013078502.2
XP_012933956.1
12 - 13
1863
67
XM_021242101.1
XP_021097760.1
11 - 12
1823
67
XM_021242102.1
XP_021097761.1
11 - 12
1743
67
XM_021242103.1
XP_021097762.1
11 - 12
1532
67
XM_021242104.1
XP_021097763.1
10 - 11
1320
67
XM_021242105.1
XP_021097764.1
11 - 12
1661
67
XM_021242106.1
XP_021097765.1
12 - 13
1754
67
XM_021242107.1
XP_021097766.1
11 - 12
1570
67
XM_021242108.1
XP_021097767.1
11 - 12
1496
67
XM_021242109.1
XP_021097768.1
11 - 12
1614
67
XM_021242110.1
XP_021097769.1
10 - 11
1501
67
XM_021242111.1
XP_021097770.1
12 - 13
1868
67
XM_021242112.1
XP_021097771.1
12 - 13
1867
67
XM_021242113.1
XP_021097772.1
12 - 13
1864
67
XM_021242115.1
XP_021097774.1
11 - 12
1742
67
XM_021242116.1
XP_021097775.1
11 - 12
1744
67
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline