Search
Search

Xdh| Gene Symbol: Xdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839257.3 | XP_004839314.1 | 8 - 9 | 936 | 59 | |
| XM_013070158.2 | XP_012925612.1 | 9 - 10 | 1022 | 59 | ||
| XM_021253694.1 | XP_021109353.1 | 8 - 9 | 936 | 59 | ||
| XM_021253695.1 | XP_021109354.1 | 8 - 9 | 936 | 59 | ||
Set Membership: |
|