Search
Search

Cacng5| Gene Symbol: Cacng5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870261.3 | XP_004870318.1 | 5 - 6 | 785 | 67 | |
| XM_013067607.2 | XP_012923061.1 | 4 - 5 | 698 | 67 | ||
| XM_013067608.2 | XP_012923062.1 | 4 - 5 | 644 | 67 | ||
Set Membership: |
|