Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hg05099422_m1
See other RBFOX1 GE Assays ›
Gene Symbol:
Rbfox1
Gene Name:
RNA binding fox-1 homolog 1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_004624824: 2997984 - 5111956 on Build HetGla_female_1.0
Species:
Heterocephalus glaber
Species Specific ID (Flybase ID):
-
Assay ID
Hg05099422_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Rbfox1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_004864846.2
XP_004864903.1
7 - 8
717
62
XM_004864848.2
XP_004864905.1
7 - 8
717
62
XM_004864850.3
XP_004864907.1
7 - 8
717
62
XM_004864857.3
XP_004864914.1
2 - 3
508
62
XM_013065582.2
XP_012921036.1
7 - 8
717
62
XM_013065583.2
XP_012921037.1
6 - 7
669
62
XM_013065584.2
XP_012921038.1
7 - 8
717
62
XM_013065585.2
XP_012921039.1
7 - 8
717
62
XM_013065586.2
XP_012921040.1
7 - 8
717
62
XM_013065587.2
XP_012921041.1
7 - 8
717
62
XM_013065588.2
XP_012921042.1
7 - 8
717
62
XM_013065589.2
XP_012921043.1
7 - 8
510
62
XM_013065590.2
XP_012921044.1
5 - 6
1225
62
XM_013065591.2
XP_012921045.1
2 - 3
651
62
XM_013065592.2
XP_012921046.1
4 - 5
496
62
XM_013065593.2
XP_012921047.1
2 - 3
509
62
XM_013065594.2
XP_012921048.1
2 - 3
545
62
XM_021244378.1
XP_021100037.1
7 - 8
717
62
XM_021244379.1
XP_021100038.1
7 - 8
717
62
XM_021244381.1
XP_021100040.1
7 - 8
717
62
XM_021244382.1
XP_021100041.1
7 - 8
717
62
XM_021244383.1
XP_021100042.1
7 - 8
717
62
XM_021244384.1
XP_021100043.1
4 - 5
983
62
XM_021244385.1
XP_021100044.1
2 - 3
321
62
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Probe spans exons
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit