Search
Search

Kansl1| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868650.3 | XP_004868707.1 | 7 - 8 | 2289 | 62 | |
| XM_004868651.2 | XP_004868708.1 | 6 - 7 | 2017 | 62 | ||
| XM_004868652.3 | XP_004868709.1 | 6 - 7 | 2188 | 62 | ||
| XM_004868653.3 | XP_004868710.1 | 7 - 8 | 2293 | 62 | ||
Set Membership: |
|