Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hg05101980_m1
          See other METTL6 GE Assays ›
          Gene Symbol:
          Mettl6
          Gene Name:
          methyltransferase like 6
          Gene Aliases:
          GW7_01596
          Chromosome Location:
          Chr.Un|NW_004624822: 5253890 - 5327234 on Build HetGla_female_1.0
          Species:
          Heterocephalus glaber
          Species Specific ID (Flybase ID):
          -
          Assay ID Hg05101980_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mettl6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_004864430.3 XP_004864487.2 4 - 5 841 89
          XM_004864431.3 XP_004864488.1 4 - 5 841 89
          XM_021244116.1 XP_021099775.1 4 - 5 841 89
          XM_021244117.1 XP_021099776.1 4 - 5 841 89
          XM_021244118.1 XP_021099777.1 4 - 5 841 89
          XM_021244119.1 XP_021099778.1 3 - 4 706 89
          XM_021244120.1 XP_021099779.1 4 - 5 841 89
          XM_021244121.1 XP_021099780.1 4 - 5 841 89
          XM_021244122.1 XP_021099781.1 4 - 5 841 89
          XM_021244123.1 XP_021099782.1 4 - 5 841 89
          XM_021244124.1 XP_021099783.1 4 - 5 841 89
          XM_021244125.1 XP_021099784.1 4 - 5 841 89
          XM_021244126.1 XP_021099785.1 4 - 5 841 89
          XM_021244127.1 XP_021099786.1 4 - 5 841 89
          XM_021244128.1 XP_021099787.1 4 - 5 841 89
          XM_021244129.1 XP_021099788.1 4 - 5 841 89
          XM_021244130.1 XP_021099789.1 4 - 5 841 89
          XM_021244131.1 XP_021099790.1 4 - 5 841 89
          XM_021244132.1 XP_021099791.1 4 - 5 841 89
          XM_021244133.1 XP_021099792.1 4 - 5 841 89
          XM_021244134.1 XP_021099793.1 4 - 5 841 89
          XM_021244135.1 XP_021099794.1 4 - 5 841 89
          XM_021244136.1 XP_021099795.1 4 - 5 841 89
          XM_021244137.1 XP_021099796.1 4 - 5 841 89
          XM_021244138.1 XP_021099797.1 4 - 5 841 89
          XM_021244139.1 XP_021099798.1 4 - 5 841 89
          XM_021244140.1 XP_021099799.1 4 - 5 841 89
          XM_021244141.1 XP_021099800.1 4 - 5 841 89
          XM_021244142.1 XP_021099801.1 4 - 5 841 89
          XM_021244144.1 XP_021099803.1 4 - 5 841 89
          XM_021244145.1 XP_021099804.1 4 - 5 841 89
          XM_021244146.1 XP_021099805.1 4 - 5 841 89
          XM_021244147.1 XP_021099806.1 4 - 5 907 89
          XM_021244148.1 XP_021099807.1 4 - 5 728 89
          XM_021244153.1 XP_021099812.1 2 - 3 542 89

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Probe spans exons

          Panther Classification:

          Molecular Function -

          methyltransferase

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline