Search
Search

Ranbp3| Gene Symbol: Ranbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865668.3 | XP_004865725.1 | 4 - 5 | 458 | 68 | |
| XM_004865671.3 | XP_004865728.1 | 3 - 4 | 257 | 68 | ||
| XM_021245153.1 | XP_021100812.1 | 4 - 5 | 459 | 68 | ||
| XM_021245154.1 | XP_021100813.1 | 3 - 4 | 257 | 68 | ||
| XM_021245155.1 | XP_021100814.1 | 4 - 5 | 460 | 68 | ||
Set Membership: |
|