Search
Search

St18| Gene Symbol: St18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837518.3 | XP_004837575.1 | 9 - 10 | 1603 | 56 | |
| XM_021248907.1 | XP_021104566.1 | 5 - 6 | 1557 | 56 | ||
| XM_021248911.1 | XP_021104570.1 | 5 - 6 | 1557 | 56 | ||
| XM_021248922.1 | XP_021104581.1 | 10 - 11 | 1659 | 56 | ||
| XM_021248923.1 | XP_021104582.1 | 11 - 12 | 1688 | 56 | ||
| XM_021248930.1 | XP_021104589.1 | 9 - 10 | 1641 | 56 | ||
| XM_021248931.1 | XP_021104590.1 | 9 - 10 | 1616 | 56 | ||
| XM_021248933.1 | XP_021104592.1 | 8 - 9 | 1560 | 56 | ||
| XM_021248937.1 | XP_021104596.1 | 8 - 9 | 1598 | 56 | ||
| XM_021248942.1 | XP_021104601.1 | 5 - 6 | 1559 | 56 | ||
Set Membership: |
|