Search
Search

Kif23| Gene Symbol: Kif23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855566.3 | XP_004855623.1 | 5 - 6 | 1525 | 65 | |
| XM_004855567.3 | XP_004855624.1 | 5 - 6 | 1529 | 65 | ||
| XM_013076268.2 | XP_012931722.1 | 5 - 6 | 1532 | 65 | ||
| XM_013076269.2 | XP_012931723.1 | 5 - 6 | 1322 | 65 | ||
Set Membership: |
|