Search
Search

Ehmt1| Gene Symbol: Ehmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848696.3 | XP_004848753.1 | 4 - 5 | 1256 | 61 | |
| XM_004848697.2 | XP_004848754.1 | 4 - 5 | 745 | 61 | ||
| XM_004848699.2 | XP_004848756.1 | 3 - 4 | 674 | 61 | ||
| XM_004848703.3 | XP_004848760.1 | 3 - 4 | 1172 | 61 | ||
| XM_004848704.3 | XP_004848761.1 | 3 - 4 | 665 | 61 | ||
| XM_004848705.3 | XP_004848762.1 | 3 - 4 | 1435 | 61 | ||
| XM_004848706.3 | XP_004848763.2 | 3 - 4 | 1198 | 61 | ||
| XM_013073666.2 | XP_012929120.1 | 3 - 4 | 1197 | 61 | ||
| XM_021261581.1 | XP_021117240.1 | 3 - 4 | 1200 | 61 | ||
| XM_021261582.1 | XP_021117241.1 | 5 - 6 | 1339 | 61 | ||
| XM_021261583.1 | XP_021117242.1 | 3 - 4 | 664 | 61 | ||
| XM_021261584.1 | XP_021117243.1 | 4 - 5 | 754 | 61 | ||
| XM_021261586.1 | XP_021117245.1 | 3 - 4 | 775 | 61 | ||
| XM_021261587.1 | XP_021117246.1 | 3 - 4 | 632 | 61 | ||
| XM_021261588.1 | XP_021117247.1 | 3 - 4 | 1199 | 61 | ||
| XM_021261589.1 | XP_021117248.1 | 3 - 4 | 1204 | 61 | ||
Set Membership: |
|