Search
Search

Tpd52l1| Gene Symbol: Tpd52l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860135.2 | XP_004860192.1 | 2 - 3 | 238 | 63 | |
| XM_004860136.2 | XP_004860193.1 | 2 - 3 | 238 | 63 | ||
| XM_004860137.3 | XP_004860194.1 | 2 - 3 | 238 | 63 | ||
| XM_013077937.2 | XP_012933391.1 | 2 - 3 | 238 | 63 | ||
| XM_021240639.1 | XP_021096298.1 | 2 - 3 | 238 | 63 | ||
| XM_021240640.1 | XP_021096299.1 | 2 - 3 | 238 | 63 | ||
| XM_021240641.1 | XP_021096300.1 | 2 - 3 | 238 | 63 | ||
Set Membership: |
|