Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hg05125709_m1
          See other FNBP1 GE Assays ›
          Gene Symbol:
          Fnbp1
          Gene Name:
          formin binding protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.Un|NW_004624760: 5488355 - 5563151 on Build HetGla_female_1.0
          Species:
          Heterocephalus glaber
          Species Specific ID (Flybase ID):
          -
          Assay ID Hg05125709_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fnbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_004849115.2 XP_004849172.1 5 - 6 612 59
          XM_004849116.2 XP_004849173.1 5 - 6 612 59
          XM_004849117.2 XP_004849174.1 5 - 6 612 59
          XM_004849118.2 XP_004849175.1 5 - 6 612 59
          XM_004849120.2 XP_004849177.1 5 - 6 612 59
          XM_004849121.2 XP_004849178.1 5 - 6 612 59
          XM_004849125.2 XP_004849182.1 5 - 6 612 59
          XM_004849126.2 XP_004849183.1 5 - 6 612 59
          XM_004849127.2 XP_004849184.1 5 - 6 612 59
          XM_004849128.3 XP_004849185.1 2 - 3 157 59
          XM_013073668.2 XP_012929122.1 5 - 6 612 59
          XM_021261871.1 XP_021117530.1 5 - 6 498 59
          XM_021261872.1 XP_021117531.1 5 - 6 612 59
          XM_021261874.1 XP_021117533.1 5 - 6 612 59
          XM_021261875.1 XP_021117534.1 5 - 6 612 59
          XM_021261876.1 XP_021117535.1 5 - 6 612 59
          XM_021261877.1 XP_021117536.1 5 - 6 415 59
          XM_021261878.1 XP_021117537.1 5 - 6 419 59
          XM_021261879.1 XP_021117538.1 5 - 6 612 59
          XM_021261880.1 XP_021117539.1 5 - 6 612 59
          XM_021261881.1 XP_021117540.1 5 - 6 612 59
          XM_021261882.1 XP_021117541.1 5 - 6 612 59
          XM_021261883.1 XP_021117542.1 5 - 6 612 59
          XM_021261884.1 XP_021117543.1 5 - 6 612 59
          XM_021261885.1 XP_021117544.1 5 - 6 612 59

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_004849128.3 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline