Search
Search

Tubgcp3| Gene Symbol: Tubgcp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858536.2 | XP_004858593.1 | 4 - 5 | 419 | 57 | |
| XM_021239303.1 | XP_021094962.1 | 4 - 5 | 419 | 57 | ||
| XM_021239304.1 | XP_021094963.1 | 4 - 5 | 419 | 57 | ||
| XM_021239305.1 | XP_021094964.1 | 2 - 3 | 178 | 57 | ||
Set Membership: |
|