Search
Search

Wls| Gene Symbol: Wls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841469.3 | XP_004841526.1 | 11 - 12 | 1718 | 68 | |
| XM_021255559.1 | XP_021111218.1 | 11 - 12 | 1754 | 68 | ||
Set Membership: |
|