Search
Search

Col24a1| Gene Symbol: Col24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841343.3 | XP_004841400.1 | 18 - 19 | 2796 | 58 | |
| XM_021255434.1 | XP_021111093.1 | 18 - 19 | 2797 | 58 | ||
| XM_021255435.1 | XP_021111094.1 | 18 - 19 | 2805 | 58 | ||
Set Membership: |
|