Search
Search

Adgb| Gene Symbol: Adgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237967.1 | XP_021093626.1 | 17 - 18 | 2287 | 73 | |
| XM_021237968.1 | XP_021093627.1 | 17 - 18 | 2286 | 73 | ||
| XM_021237969.1 | XP_021093628.1 | 17 - 18 | 2288 | 73 | ||
| XM_021237970.1 | XP_021093629.1 | 16 - 17 | 2077 | 73 | ||
| XM_021237971.1 | XP_021093630.1 | 16 - 17 | 2046 | 73 | ||
| XM_021237972.1 | XP_021093631.1 | 14 - 15 | 1697 | 73 | ||
| XM_021237973.1 | XP_021093632.1 | 11 - 12 | 1343 | 73 | ||
Set Membership: |
|