Search
Search

Znf84| Gene Symbol: Znf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844042.3 | XP_004844099.1 | 5 - 6 | 609 | 76 | |
| XM_021257742.1 | XP_021113401.1 | 5 - 6 | 706 | 76 | ||
| XM_021257743.1 | XP_021113402.1 | 5 - 6 | 718 | 76 | ||
| XM_021257744.1 | XP_021113403.1 | 5 - 6 | 650 | 76 | ||
| XM_021257745.1 | XP_021113404.1 | 5 - 6 | 928 | 76 | ||
| XM_021257746.1 | XP_021113405.1 | 4 - 5 | 483 | 76 | ||
Set Membership: |
|