Search
Search

Rab40c| Gene Symbol: Rab40c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873790.3 | XP_004873847.1 | 3 - 4 | 269 | 72 | |
| XM_021251442.1 | XP_021107101.1 | 3 - 4 | 267 | 72 | ||
| XM_021251443.1 | XP_021107102.1 | 3 - 4 | 499 | 72 | ||
| XM_021251444.1 | XP_021107103.1 | 2 - 3 | 187 | 72 | ||
Set Membership: |
|