Search
Search

Fbxo45| Gene Symbol: Fbxo45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834700.2 | XP_004834757.1 | 1 - 2 | 442 | 65 | |
| XM_021255087.1 | XP_021110746.1 | 1 - 2 | 442 | 65 | ||
Set Membership: |
|