Search
Search

Celsr2| Gene Symbol: Celsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853622.3 | XP_004853679.1 | 19 - 20 | 6683 | 55 | |
| XM_013075541.2 | XP_012930995.1 | 19 - 20 | 6683 | 55 | ||
Set Membership: |
|