Search
Search

Ptpn1| Gene Symbol: Ptpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077154.2 | XP_012932608.1 | 7 - 8 | 1043 | 62 | |
| XM_013077155.2 | XP_012932609.1 | 7 - 8 | 1043 | 62 | ||
| XM_021238924.1 | XP_021094583.1 | 5 - 6 | 851 | 62 | ||
Set Membership: |
|