Search
Search

Asmtl| Gene Symbol: Asmtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867313.3 | XP_004867370.1 | 10 - 11 | 1424 | 98 | |
| XM_021246366.1 | XP_021102025.1 | 10 - 11 | 1931 | 98 | ||
Set Membership: |
|