Search
Search

Znf18| Gene Symbol: Znf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857550.3 | XP_004857607.1 | 3 - 4 | 1463 | 77 | |
| XM_004857551.3 | XP_004857608.1 | 3 - 4 | 739 | 77 | ||
| XM_021238164.1 | XP_021093823.1 | 5 - 6 | 1048 | 77 | ||
| XM_021238165.1 | XP_021093824.1 | 4 - 5 | 936 | 77 | ||
| XM_021238166.1 | XP_021093825.1 | 5 - 6 | 1030 | 77 | ||
| XM_021238168.1 | XP_021093827.1 | 4 - 5 | 918 | 77 | ||
| XM_021238169.1 | XP_021093828.1 | 4 - 5 | 1052 | 77 | ||
| XM_021238170.1 | XP_021093829.1 | 4 - 5 | 895 | 77 | ||
| XM_021238171.1 | XP_021093830.1 | 3 - 4 | 794 | 77 | ||
| XM_021238172.1 | XP_021093831.1 | 3 - 4 | 788 | 77 | ||
| XM_021238173.1 | XP_021093832.1 | 3 - 4 | 806 | 77 | ||
| XM_021238174.1 | XP_021093833.1 | 2 - 3 | 579 | 77 | ||
| XM_021238175.1 | XP_021093834.1 | 3 - 4 | 1463 | 77 | ||
| XM_021238176.1 | XP_021093835.1 | 3 - 4 | 1463 | 77 | ||
| XM_021238177.1 | XP_021093836.1 | 3 - 4 | 1463 | 77 | ||
| XM_021238178.1 | XP_021093837.1 | 3 - 4 | 1463 | 77 | ||
Set Membership: |
|