Search
Search

Ano5| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013074705.2 | XP_012930159.1 | 18 - 19 | 2381 | 60 | |
| XM_013074706.2 | XP_012930160.1 | 17 - 18 | 2333 | 60 | ||
| XM_021263924.1 | XP_021119583.1 | 19 - 20 | 2423 | 60 | ||
| XM_021263925.1 | XP_021119584.1 | 19 - 20 | 2444 | 60 | ||
| XM_021263926.1 | XP_021119585.1 | 19 - 20 | 2423 | 60 | ||
| XM_021263927.1 | XP_021119586.1 | 17 - 18 | 2288 | 60 | ||
| XM_021263928.1 | XP_021119587.1 | 14 - 15 | 1919 | 60 | ||
| XM_021263929.1 | XP_021119588.1 | 15 - 16 | 1924 | 60 | ||
Set Membership: |
|