Search
Search

Spaca6| Gene Symbol: Spaca6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066323.2 | XP_012921777.1 | 8 - 9 | 1111 | 91 | |
| XM_021246273.1 | XP_021101932.1 | 8 - 9 | 1144 | 91 | ||
| XM_021246274.1 | XP_021101933.1 | 8 - 9 | 1106 | 91 | ||
| XM_021246275.1 | XP_021101934.1 | 8 - 9 | 1063 | 91 | ||
Set Membership: |
|