Search
Search

Synrg| Gene Symbol: Synrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870422.3 | XP_004870479.1 | 14 - 15 | 2991 | 61 | |
| XM_013067672.2 | XP_012923126.1 | 16 - 17 | 3641 | 61 | ||
| XM_013067673.2 | XP_012923127.1 | 16 - 17 | 3528 | 61 | ||
| XM_013067674.2 | XP_012923128.1 | 15 - 16 | 3465 | 61 | ||
| XM_013067675.2 | XP_012923129.1 | 16 - 17 | 3435 | 61 | ||
| XM_013067676.2 | XP_012923130.1 | 16 - 17 | 3390 | 61 | ||
| XM_013067677.2 | XP_012923131.1 | 15 - 16 | 3294 | 61 | ||
| XM_013067678.2 | XP_012923132.1 | 15 - 16 | 3201 | 61 | ||
| XM_021248720.1 | XP_021104379.1 | 16 - 17 | 3620 | 61 | ||
| XM_021248721.1 | XP_021104380.1 | 16 - 17 | 3393 | 61 | ||
| XM_021248722.1 | XP_021104381.1 | 13 - 14 | 3265 | 61 | ||
| XM_021248723.1 | XP_021104382.1 | 14 - 15 | 2991 | 61 | ||
| XM_021248724.1 | XP_021104383.1 | 14 - 15 | 2856 | 61 | ||
| XM_021248725.1 | XP_021104384.1 | 14 - 15 | 2856 | 61 | ||
Set Membership: |
|