Search
Search

Fgr| Gene Symbol: Fgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850726.3 | XP_004850783.1 | 5 - 6 | 661 | 61 | |
| XM_004850727.3 | XP_004850784.1 | 5 - 6 | 689 | 61 | ||
| XM_013074392.2 | XP_012929846.1 | 5 - 6 | 686 | 61 | ||
| XM_021263158.1 | XP_021118817.1 | 5 - 6 | 658 | 61 | ||
| XM_021263159.1 | XP_021118818.1 | 5 - 6 | 691 | 61 | ||
Set Membership: |
|