Search
Search

Nudt17| Gene Symbol: Nudt17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853912.3 | XP_004853969.1 | 6 - 7 | 787 | 98 | |
| XM_021265818.1 | XP_021121477.1 | 5 - 6 | 922 | 98 | ||
| XM_021265821.1 | XP_021121480.1 | 5 - 6 | 814 | 98 | ||
Set Membership: |
|