Search
Search

Sash1| Gene Symbol: Sash1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856903.2 | XP_004856960.1 | 13 - 14 | 1530 | 73 | |
| XM_021238001.1 | XP_021093660.1 | 13 - 14 | 2058 | 73 | ||
| XM_021238002.1 | XP_021093661.1 | 13 - 14 | 1924 | 73 | ||
| XM_021238003.1 | XP_021093662.1 | 6 - 7 | 1730 | 73 | ||
| XM_021238004.1 | XP_021093663.1 | 5 - 6 | 852 | 73 | ||
Set Membership: |
|