Search
Search

Slc44a5| Gene Symbol: Slc44a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255525.1 | XP_021111184.1 | 11 - 12 | 777 | 63 | |
| XM_021255526.1 | XP_021111185.1 | 11 - 12 | 767 | 63 | ||
| XM_021255527.1 | XP_021111186.1 | 10 - 11 | 689 | 63 | ||
| XM_021255528.1 | XP_021111187.1 | 11 - 12 | 777 | 63 | ||
| XM_021255529.1 | XP_021111188.1 | 10 - 11 | 738 | 63 | ||
| XM_021255530.1 | XP_021111189.1 | 10 - 11 | 728 | 63 | ||
| XM_021255531.1 | XP_021111190.1 | 9 - 10 | 650 | 63 | ||
| XM_021255532.1 | XP_021111191.1 | 7 - 8 | 566 | 63 | ||
Set Membership: |
|