Search
Search

Pcnx1| Gene Symbol: Pcnx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837289.2 | XP_004837346.1 | 23 - 24 | 4751 | 62 | |
| XM_013067460.2 | XP_012922914.1 | 23 - 24 | 4748 | 62 | ||
| XM_013067463.2 | XP_012922917.1 | 23 - 24 | 4736 | 62 | ||
| XM_013067464.2 | XP_012922918.1 | 21 - 22 | 4433 | 62 | ||
| XM_013067465.2 | XP_012922919.1 | 21 - 22 | 4418 | 62 | ||
| XM_021247065.1 | XP_021102724.1 | 23 - 24 | 4752 | 62 | ||
| XM_021247069.1 | XP_021102728.1 | 23 - 24 | 4323 | 62 | ||
| XM_021247077.1 | XP_021102736.1 | 22 - 23 | 4131 | 62 | ||
Set Membership: |
|