Search
Search

B4galnt1| Gene Symbol: B4galnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861187.3 | XP_004861244.1 | 4 - 5 | 478 | 58 | |
| XM_004861189.3 | XP_004861246.1 | 3 - 4 | 699 | 58 | ||
Set Membership: |
|