Search
Search

Catsper3| Gene Symbol: Catsper3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836218.2 | XP_004836275.1 | 6 - 7 | 985 | 99 | |
| XM_021238600.1 | XP_021094259.1 | 5 - 6 | 802 | 99 | ||
Set Membership: |
|