Search
Search

Dnah9| Gene Symbol: Dnah9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857554.2 | XP_004857611.1 | 41 - 42 | 8102 | 150 | |
| XM_021238161.1 | XP_021093820.1 | 41 - 42 | 8102 | 150 | ||
| XM_021238162.1 | XP_021093821.1 | 41 - 42 | 8104 | 150 | ||
Set Membership: |
|