Search
Search

Ezh2| Gene Symbol: Ezh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860469.3 | XP_004860526.1 | 17 - 18 | 2124 | 58 | |
| XM_004860471.3 | XP_004860528.1 | 17 - 18 | 2195 | 58 | ||
| XM_021240895.1 | XP_021096554.1 | 17 - 18 | 2286 | 58 | ||
| XM_021240896.1 | XP_021096555.1 | 17 - 18 | 2146 | 58 | ||
| XM_021240897.1 | XP_021096556.1 | 17 - 18 | 2060 | 58 | ||
| XM_021240898.1 | XP_021096557.1 | 17 - 18 | 2109 | 58 | ||
| XM_021240899.1 | XP_021096558.1 | 17 - 18 | 2097 | 58 | ||
| XM_021240900.1 | XP_021096559.1 | 17 - 18 | 2082 | 58 | ||
| XM_021240901.1 | XP_021096560.1 | 17 - 18 | 2153 | 58 | ||
Set Membership: |
|